missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BMP5 Recombinant Human Protein, N-hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the carboxy-terminal domain (Gln 324-His 454) (the mature chain) of human BMP5 (NP_066551.1) fused to Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10214H01H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15224935

  • £653.75 / 50µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

Bone Morphogenetic Proteins 5 (BMP5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
653
PAGE
50μg
N-Terminal
<1 EU/μg
Bone morphogenetic proteins 5
Assay Standard
Mammalian
Lyophilized
41.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
BMP5, mgC34244
BMP5
Growth Factors & Receptors
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.