missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BLMH Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human BLMH (NP_000377.1) (Ser 2-Glu 455) fused to a polyhistidinetag at the N-terminus

Brand:  Thermo Scientific™ 11333H07E25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15278535

  • £415.36 / 25ยตg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Bleomycin hydrolase (BLMH), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications., or in other research applications.
  • Activity: Measured by its ability to hydrolyze Met-AMC; the specific activity is >500pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
53.4kDa
5 x 5μg
Proteases
Bleomycin hydrolase
Assay Standard
E. coli
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
NME1, AWD, GAAD, NB, NBS, NDPK-A, NDPKA, nM23, nM23-H1
BLMH
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.