Sino Biological™ BCL2L2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10059H08E25

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15224575

Quantity Price
1 £ 121.0 / 25µg
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype Other Proteins
Storage Requirements Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5 x 5μg
Purification Method PAGE
Gene ID (Entrez) 599
Validated Application Assay Standard
Common Name Bcl2-Like 2
Protein Family Other Proteins
Protein Tag His Tag
Structural Form Full Length/Recombinant
Expression System E. coli
Purity ≥90% by SDS-PAGE
Gene Alias BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
Gene Symbol BCL2L2
Molecular Weight 20kDa
Format Frozen

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 25mM HEPES, 100mM KCl, 10% Glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.