missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ BCHE Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse BCHE (NP_033868.3) (Met 1-Leu 603) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 50418M08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15383206

  • £142.62 / 5µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

Description

Description

Butyrylcholinesterase (BCHE), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave butyrylthiocholine; the specific activity is >300nM/min./μg
  • Formulation: Lyophilized in 25mM Tris, 100mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
66.7kDa
5μg
Glycoproteins
C730038G20Rik, mgC107651, Bche
BCHE
Other Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Butyrylcholinesterase
Assay Standard
Mammalian
Videos