missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE1 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 457) of the mouse BACE1 precursor (NM_011792.5) fused to the polyhistidine tag at the C-terminus
£58.45 - £230.00
Specifications
For Use With (Application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories |
---|---|
Molecular Weight (g/mol) | 49.8kDa |
Storage Requirements | Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. |
Endotoxin Concentration | <1 EU/μg |
Gene Alias | Bace1, C76936 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15302516
|
Invitrogen
50002M08H5 |
5μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15312516
|
Invitrogen
50002M08H50 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Beta-site APP-cleaving enzyme 1 (BACE1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to cleave a fluorescent peptide substrate Mca-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(Dpn)-Arg-Arg-NH2, cleavage of this substrate can be measured using excitation and emission wavelengths of 320 and 405nM, respectively; the specific activity is >2pmole/min./μg
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Bace1, C76936 | |
BACE1 | |
Mammalian | |
Lyophilized | |
≥90% by SDS-PAGE |
49.8kDa | |
<1 EU/μg | |
beta-site APP cleaving enzyme 1 | |
His Tag | |
Other Proteins | |
Proteases |
For Research Use Only. Not for use in diagnostic procedures.