missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ B2M Recombinant Mouse Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse B2M (P01887) (Met 1-Met 119), without the propeptide, fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 50957M08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15309336

  • £203.00 / 25µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Description

Description

Beta-2-microglobulin (B2M), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12010
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Beta-2-microglobulin
Assay Standard
Mammalian
Lyophilized
13kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
RP23-34E24.5, Ly-m11, beta2-m, beta2m
B2M
MHC (Major Histocompatibility Complex) Proteins
His Tag
Videos