missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ APP Recombinant Human Protein, N-His.GST Tag

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Asp 672-Val 711) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 40, fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Brand:  Thermo Scientific™ 10703H20E15

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15247105

  • £54.92 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Amyloid beta (A4) precursor (APP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated recombinant human AGER (Mfr. No. 11629-HCCH) in a functional ELISA
  • Formulation: Lyophilized in 50mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
351
PAGE
5μg
N-Terminal
<1 EU/μg
Amyloid beta (A4) precursor protein
Assay Standard
E. coli
Lyophilized
31.8kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
Beta-APP40
APP
Other Proteins
GST Tag, His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.