missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ APOE Recombinant Human Protein, N-TRX.His Tag

This recombinant protein was expressed from a DNA sequence encoding the human APOE (P02649) (Lys 19-His 317) fused to a Trx and a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10817H30E5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227235

  • £37.40 / 5µg


Estimated Shipment
12-06-2018
Add to basket

Description

Description

Apolipoprotein E (APOE), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: PBS, 40% Glycerol, 0.5mM TCEP, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
348
PAGE
5μg
Full Length/Recombinant
AD2, LDLCQ5, LPG, mgC1571
APOE
Other Proteins
Thioredoxin, His Tag
Frozen
51.3kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
Apolipoprotein E
Assay Standard
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.