missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ANXA5 Recombinant Human Protein

This recombinant protein was expressed from a DNA sequence encoding the human ANXA5 (P08758) (Met 1-Asp 320)

Brand:  Thermo Scientific™ 10448HNAE50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15295435

  • £186.34 / 50µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Annexin A5 (ANXA5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 240mM NaCl, pH 8.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
308
PAGE
50μg
Full Length/Recombinant
ANX5, ENX2, PP4
ANXA5
Other Proteins
Lyophilized
35.9kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
<1 EU/μg
Annexin A5
Assay Standard
E. coli
Videos