missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ANPEP Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the metalloprotease domain (Lys 69-Lys 967, 603 Met/Ile) of the mature form of human ANPEP (NP_001141.2) with the signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10051H08H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15224555

  • £2748.24 / 250µg


Estimated Shipment
16-05-2018
Add to basket

Description

Description

Alanyl Amonipeptidase (ANPEP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Ala-7-amido-4-methylcoumarin (Ala-AMC); the specific activity is >2500 pmoles/min./μg
  • Formulation: Lyophilized in 25mM MES, 0.15M NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
104.2kDa
5 x 50μg
Proteases
ANPEP, APN, CD13, LAP1, PEPN, gp150
ANPEP
CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Alanyl (membrane) aminopeptidase
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.