missing translation for 'onlineSavingsMsg'
Learn More

ANGPTL5 Recombinant Human Protein, GST Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 26-Lys 388) of human ANGPTL5 (NP_835228.2) fused to a GST tag at the N-terminus

Brand:  Invitrogen 11094H09B25

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 13486096

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Angiopoietin-like Protein 5 (ANGPTL5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 0.5mM PMSF, 1mM reduced Glutathione, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
68kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Angiopoietin-like 5
GST Tag
Growth Factors & Receptors
Growth Factor Receptors
253935
5 x 5μg
Recombinant
ANGPTL5
ANGPTL5
Insect
Lyophilized
≥90% by SDS-PAGE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.