missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ANGPT2 (ANG2) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human ANGPT2 (NP_001138.1) (Met 1-Phe 496) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10691H08H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15287055

  • £205.08 / 25µg

EU Stock 2
Add to basket



Angiopoietin 2 (ANGPT2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind recombinant human Tie2-His in a functional ELISA; 2. Measured by its ability to bind recombinant human Tie2-Fch in a functional ELISA
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.



Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
5 x 5μg
Other Growth Factors
Growth Factors & Receptors
His Tag
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Angiopoietin 2
Assay Standard

For Research Use Only. Not for use in diagnostic procedures.