missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ALPL Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human tissue-nonspecific alkaline phosphatase (NP_000469.3) (Met 1-Ser 502) fused to a C-terminal polyhistidine tag

Brand:  Thermo Scientific™ 10440H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15205425

  • £346.66 / 25µg


Estimated Shipment
09-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Alkaline Phosphatase, liver/bone/kidney (ALPL), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 25mM Tris, 0.15M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
249
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Alkaline Phosphatase, liver/bone/kidney
Assay Standard
Mammalian
Lyophilized
55kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
ALPL, AP-TNAP, FLJ40094, FLJ93059, HOPS, mgC161443, mgC167935, TNAP, TNSALP
ALPL
Other Proteins
His Tag
Videos