missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ALDH1A1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human ALDH1A1 (NP_000680.2) (Ser 2-Ser 501) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11388H07E25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15288555

  • £260.25 / 25µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
216
PAGE
5 x 5μg
N-Terminal
<1 EU/μg
Aldehyde dehydrogenase 1 family, member A1
Assay Standard
E. coli
Lyophilized
41.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
ALDH1A1, RP11-151D14.2, ALDC, ALDH-E1, ALDH1, ALDH11, mgC2318, PUMB1, RALDH1
ALDH1A1
Other Proteins
His Tag
Videos