Sino Biological™ ALCAM (CD166) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10045H08H5

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13485666

Quantity Price
1 £ 54.75 / 5µg
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Specification

Expression System Mammalian
Protein Tag His Tag
Gene ID (Entrez) 214
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Family CD Proteins
Quantity 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Common Name Activated leukocyte cell adhesion molecule
Gene Symbol ALCAM
Protein Subtype Glycoproteins
Molecular Weight 57.4kDa
Endotoxin Concentration <1 EU/μg
Gene Alias CD166, FLJ38514, MEMD, mgC71733, ALCAM
Validated Application Assay Standard
Format Lyophilized

Activated leukocyte cell adhesion molecule (ALCAM), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.