missing translation for 'onlineSavingsMsg'
Learn More

ALCAM (CD166) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 10045H08H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465666

  • £147.00 / 25µg


Estimated Shipment
10-04-2019
Add to basket

Description

Description

Activated leukocyte cell adhesion molecule (ALCAM), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
214
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD166, FLJ38514, MEMD, mgC71733, ALCAM
ALCAM
CD Proteins
His Tag
Lyophilized
57.4kDa
5 x 5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Activated leukocyte cell adhesion molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.