missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ALCAM (CD166) Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10045H03H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15294535

  • £187.38 / 50µg


Estimated Shipment
09-05-2018
Add to basket

Description

Description

Activated leukocyte cell adhesion molecule (ALCAM), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
214
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD166, FLJ38514, MEMD, mgC71733, ALCAM
ALCAM
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
84kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Activated leukocyte cell adhesion molecule
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.