missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ AKR1B1 Recombinant Human Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the human AKR1B1 (P15121) (Met 1-Phe 316) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 11294H07E250

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208495

  • £519.46 / 250µg


Estimated Shipment
14-05-2018
Add to basket

Description

Description

Aldo-Keto Reductase family 1 member B1 (aldose reductase) (AKR1B1), recombinant human protein is supplied frozen. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: PBS, 20% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
231
PAGE
5 x 5μg
Full Length/Recombinant
Aldo-Keto Reductase Family 1, Member B1 (Aldose Reductase)
Assay Standard
E. coli
Frozen
37.9kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ADR, ALDR1, ALR2, AR, mgC1804
AKR1B1
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.