missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ AIM2 Recombinant Human Protein, N-GST tag

This recombinant protein was expressed from a DNA sequence encoding the human AIM2 (NP_004824.1) (Met 1-Thr 343) fused to the GST tag at the N-terminus

Brand:  Thermo Scientific™ 11654H09B50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15278785

  • £333.12 / 50µg


Estimated Shipment
06-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Absent In Melanoma 2 (AIM2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells (Karey, K.P. et al. (1988) Cancer Research 48:4083); the ED50 for this effect is typically 4-7μg/mL
  • Formulation: Lyophilized in 50mM Tris, 1M NaCl, 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
65.2kDa
50μg
NFκB Pathway Proteins
AIM2, RP11-520H16.3, PYHIN4
AIM2
Other Proteins
GST Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Absent in melanoma 2
Assay Standard
Insect
Videos