missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ AGER Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AGER isoform 1 (NP_001127.1) (Met 1-Ala 344) fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 11629H02H25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15278745

  • £123.88 / 25µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Advanced glycosylation end product-specific receptor (AGER), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human S100A12 at 2μg/mL (100μL/well) can bind recombinant human AGER with a linear range of 0.032-20μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
61.1kDa
5 x 5μg
Other Proteins
DAMA-358M23.4, mgC22357, RAGE
AGER
Other Proteins
IgG1 Fc Tag
Lyophilized
>70% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Advanced glycosylation end product-specific receptor
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.