missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ACVR2B Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 134) of mouse ACVR2B (NP_031423.1) precursor fused to the Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 50173M03H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15332866

  • £234.23 / 50µg


Estimated Shipment
05-07-2018
Add to basket

Includes: One vial of protein in suspension buffer

Description

Description

Activin Receptor IIB (ACVR2B), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: 1. Measured by its ability to bind biotinylated Human INHBA-his (Mfr. No. 10429-H08H) in functional ELISA; 2. Measured by its ability to bind biotinylated mouse INHBA-his (Mfr. No. 50659-M08H) in functional ELISA
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Growth Factors & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.