missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ACVR1B (ALK4) Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc His Tag

This recombinant protein was expressed from a DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) fused to C-terminal His-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10583H03H50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15216965

  • £165.52 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

Activin A Receptor, Type Ib (ACVR1B), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human TDGF1 at 2μg/mL (100μL/well) can bind human ACVR1B with a linear range of 0.0068-0.16μg/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Growth Factors & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.