missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ ACP1 Recombinant Human Protein, GST Tag

This recombinant protein was expressed from a DNA sequence encoding human ACP1 (AAI06012.1) (Met 1-His 158) fused to a GST tag at the N-terminus

Brand:  Thermo Scientific™ 10957H09E250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15267445

  • £655.83 / 250µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Acid phosphatase 1, soluble (ACP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave a substrate, pNitrophenyl phosphate (pNPP), the specific activity is >65nmol/min./μg
  • Formulation: Lyophilized in 50mM Tris, 150mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
44.3kDa
5 x 50μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
HAAP, mgC111030, mgC3499
ACP1
Other Proteins
GST Tag
Lyophilized
≥85% by SDS-PAGE
Human
Other Proteins
Acid phosphatase 1, soluble
Assay Standard
E. coli
Videos