missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ A2M Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human A2M (NP_000005.2) (Met 1-Ala 1474) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10952H08B5

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15247445

  • £83.81 / 5µg


Estimated Shipment
09-07-2018
Add to basket

Description

Description

Alpha-2-Macroglobulin (A2M), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to trap trypsin; the trapped trypsin is no longer able to interact with protein substrates or inhibitors, but still able to cleave small peptide substrates or inhibitors, the IC50 value is <5nM
  • Formulation: 20mM Tris, 500mM NaCl, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
164kDa
5μg
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
alpha-2-macroglobulin
Cell Structure/Organelle Proteins/Phosphatase Inhibitors
His Tag
Frozen
≥90% by SDS-PAGE
Human
Protease Inhibitors
A2mp
A2M
Insect
Videos

For Research Use Only. Not for use in diagnostic procedures.