missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Magnetic Stir Bar, Single-sided crosshead

Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on one side of the stirrer.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.04324kg

 View more versions of this product

Product Code. 10690443

  • £8.95 / Each
Estimated Shipment: 22-04-2024
to see stock.
Add to basket

Description

Description

  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Autoclavable
Specifications

Specifications

40 mm
17mm
2/Pk.
Yes
PTFE encapsulated Alnico V magnet core
White
Containers & Vessels / Stirrer Hot Plates
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers