Learn More
Singe-Use Calibration Buffers, Color-Coded
Can be used for direct pH electrode calibration and then discarded.
Brand: Reagecon MXC65
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Color-coded by pH
- Mixed pack of 6 x 90 mL packages for pH electrode calibration at 25°C
- Tested and Certified by Reagecon’s ISO 17025 Accredited Test Method.
- Convenience - saves time, more efficient calibration, avoids waste and spillage.
- Mobility - These are easy to store and transport, allowing calibration in the field or directly in the plant.
- Economical - No waste buffer, beaker not required.
- Accuracy - the possibility of contamination is eliminated giving increased confidence in the results.
- Traceability - Each container is labelled with lot number and expiry date and buffers are directly traceable to N.I.S.T. Standards
Specifications
Red, Yellow, Blue | |
4, 7, 10 ±0.01 | |
6 x 90 mL |
Safety and Handling
- BUFFER SOLUTION pH 4.00 / 7.00 / 10.00 ± 0.01 @25ºC Certified Traceable to N.I.S.T.
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid