Learn More
Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
For use with Thermo Scientific™ Shandon Cytospin 4 Centrifuges
POA
Specifications
For Use With (Equipment) Shandon Cytospin™ 4 Cytocentrifuge
View More Specs
Description
Description
  • All plastic disposable sample chambers do not need metal clips
  • Available with or without pre-attached filter card
Specifications
Specifications
Shandon Cytospin™ 4 Cytocentrifuge