Simport™ Scientific Capinserts™

Made of polypropylene

Overview
Brand: Simport™ Scientific

Manufacturer Part Number: T345B

X500 Cap insert Fisherbrand polypropylene blue

UNSPSC: 41103913

Code: 46

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11651360

Quantity Price
1 £ 14.65 / Pack of 500
In Stock 1
Add to basket


Description and Specification

Specification

For Use With (Equipment) Microtube microcentrifuge tubes and other products
Color Blue
Material Polypropylene
For Use With (Application) Used to color code and write on frosted area for sample identification
Documentation