Simport™ Scientific Capinserts™

Made of polypropylene

Overview
Brand: Simport™ Scientific

Manufacturer Part Number: T345R

UNSPSC: 41103913

Code: 46

Additional Details:
Additional Details: Weight: 2.72160kg



Product Code. 11611370

Quantity Price
1 £ 14.85 / Pack of 500
Estimated Shipment
11-04-2017
Add to basket


Description and Specification

Specification

For Use With (Equipment) Microtube microcentrifuge tubes and other products
Material Polypropylene
Color Red
For Use With (Application) Used to color code and write on frosted area for sample identification
Documentation