Learn More
Simport Scientific Capinsert™ Cap Inserts
Made of polypropylene.
Brand:  Simport Scientific T345AS
Code : 46
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11641360
  • £14.55 / Pack of 500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
The Capinsert™ is used to color code a multitude of products such as tubes and vials. It is inserted on top of the closure and has a write-on frosted area for sample identification.
  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings
Order Info
Available in 11 colors; assorted packages contain 100 of each color
Specifications
Specifications
Yes
Polypropylene
Micrewtubes microcentrifuge tubes and other products
No
Microtube microcentrifuge tubes and other products
Assorted:Blue,lilac,red,yellow and white
Assorted
No
Used to color code and write on frosted area for sample identification
Polypropylene
Videos
Documentation