Learn More
Simport Scientific Capinsert™ Cap Inserts
Made of polypropylene.
Brand:  Simport Scientific T345G
Code : 46
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11661360
  • £14.95 / Pack of 500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
The Capinsert™ is used to color code a multitude of products such as tubes and vials. It is inserted on top of the closure and has a write-on frosted area for sample identification.
  • Simplifies sample identification
  • Press into place
  • Frosted writing area on top provides space for additional identifying markings
Order Info
Available in 11 colors; assorted packages contain 100 of each color
Specifications
Specifications
Yes
Polypropylene
Micrewtubes microcentrifuge tubes and other products
No
Microtube microcentrifuge tubes and other products
Green
Green
No
Used to color code and write on frosted area for sample identification
Polypropylene
Videos
Documentation