Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
For holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics
Brand:  Simport™ Scientific M480-2
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10569311
  • £99.65 / Pack of 1500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Made of acetal polymer and resistant to chemical action of histological solvents
  • Fit securely in microtome chuck adapters
  • Accepts standard metal lids that keep specimens safe during processing
  • Slanted writing surface for easy sample identification
  • Available in three colors
Specifications
Specifications
White
Acetal polymer
Embedding cassette
Videos