missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 99.95% (metals basis), Alfa Aesar™

Brand:  Alfa Aesar 045071.G6

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg

Quantity 10m

 View more versions of this product

Product Code. 15486715

  • £158.40 / 10m


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

Silver
7440-22-4
99.95% (Metals basis)
Hard
Wire, 0.203mm (0.008 in.) dia.
MFCD00003397

Specifications

Silver
Hard
Silver-colored wire
Wire, 0.203mm (0.008 in.) dia.
MFCD00003397
Silver
10m
7440-22-4
10.5g/cm3
99.95% (Metals basis)
Characteristic
Videos
SDS