missing translation for 'onlineSavingsMsg'
Learn More

Silver wire, 1.0mm (0.04in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4782133
Click to view available options
Quantity:
1 m
25 m
5 m
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

Form: Approx.8.24g/m; Temper: hard

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored wire
Assay Percent Range 99.9% (Metals basis)
Quantity 1 m
Odor Characteristic
Form Wire
Chemical Name or Material Silver, Hard

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'