missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver wire, 0.015mm (0.0006 in.) dia., hard, 99.99% (metals basis)

Brand:  Alfa Aesar™ 040732.G1

UNSPSC : 12141740

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg

Quantity 1m

 View more versions of this product

Product Code. 11358658

  • £371.53 / 1m


Estimated Shipment date
from Supplier 07-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Silver wire
7440-22-4
0.015mm (0.0006 in.) dia., Hard
MFCD00003397

Specifications

Silver wire
1m
7440-22-4
99.99%
MFCD00003397
≈0.0019g/m
0.015mm (0.0006 in.) dia., Hard
Wire
(metals basis)
Characteristic
Documentation

Technical Documents