missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver wire, 0.015mm (0.0006 in.) dia., annealed, 99.99% (metals basis)

Microelectronics, contacts and coatings

Brand:  Alfa Aesar™ 040731.G1

UNSPSC : 12141740

Code : NEW

Additional Details : CAS Number : 7440-22-4 Weight : 0.01000kg

Quantity 1m

 View more versions of this product

Product Code. 11328658

  • £309.41 / 1m


Estimated Shipment date
from Supplier 12-06-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Silver wire
7440-22-4
0.015mm (0.0006 in.) dia., Annealed, Temper: soft
MFCD00003397

Specifications

MFCD00003397
99.99%
7440-22-4
≈0.0019g/m
1m
(metals basis)
Silver wire
0.015mm (0.0006 in.) dia., Annealed, Temper: soft
Characteristic
Wire
Documentation

Technical Documents