missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Silver tungsten oxide, 99%

Brand:  Alfa Aesar™ 039661.09

Code : NEW

Additional Details : CAS Number : 13465-93-5 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11304617

  • £61.80 / 10g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Silver tungsten oxide
Ag2WO4
MFCD00014147
13465-93-5
463.58
Silver tungstate

Specifications

Silver tungsten oxide
13465-93-5
99%
463.58
Silver tungstate
Odorless
Light yellow
10g
Powder
Ag2WO4
MFCD00014147
Insoluble in water; Soluble in potassium cyanide, ammonium hydroxide and nitric acid
Light Sensitive
Videos
SDS