Silver trifluoromethanesulfonate, 99+%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 175200250

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 2923-28-6 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 10701071

Quantity Price
1 £ 115.0 / 25g
Estimated Shipment date
from Supplier 16-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Silver trifluoromethanesulfonate
Assay Percent Range 99+%
Molecular Formula CAgF3O3S
Linear Formula CF3SO3Ag
Formula Weight 256.92
Synonym Silver triflate

Specification

Chemical Name or Material Silver trifluoromethanesulfonate
CAS 2923-28-6
Health Hazard 1 Danger
Physical Form Powder
Elemental Analysis 99.0% min.
Assay Percent Range 99+%
Molecular Formula CAgF3O3S
Infrared Spectrum Authentic
Linear Formula CF3SO3Ag
Formula Weight 256.92
Synonym Silver triflate
Packaging Glass bottle
Color White to light beige
Quantity 25g
Documentation

Technical documents

Safety and Handling

Danger