missing translation for 'onlineSavingsMsg'
Learn More

Silver trifluoroacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-4412861
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2966-50-9
C2AgF3O2
220.883
MFCD00013199
KZJPVUDYAMEDRM-UHFFFAOYSA-M
silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
76299
silver;2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)[O-].[Ag+]
This item is not returnable. View return policy
2966-50-9
C2AgF3O2
220.883
MFCD00013199
KZJPVUDYAMEDRM-UHFFFAOYSA-M
silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
76299
silver;2,2,2-trifluoroacetate
C(=O)(C(F)(F)F)[O-].[Ag+]

Silver trifluoroacetate is used as a Lewis acid catalyst. It is used as an important raw material for the preparation of triethylsilyl trifloroacetate and triphenylmethyl trifluoroacetate. Further, it is used as a catalyst in the preparation of 4-iodoveratrole from veratrole by reacting with iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 257°C to 260°C (decomposition)
Physical Form Powder
Quantity 5 g
Assay Percent Range 98%
Solubility Information Soluble in water,ether and benzene.
Formula Weight 220.88
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Silver trifluoroacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'