missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver thiocyanate, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Silver thiocyanate is applied in the preparation of nonmetallic thiocyanates, analytical reagent, in photographic emulsions and as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 10 g |
Solubility Information | Very slightly soluble in water. Insoluble in ethanol,acetone,acid. Soluble in NH4OH |
Formula Weight | 165.95 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | White crystals |
Chemical Name or Material | Silver thiocyanate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'