missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfide, 99%, Thermo Scientific Chemicals

Catalog Number p-4407830
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
21548-73-2
Ag2S
247.80
MFCD00003406
XUARKZBEFFVFRG-UHFFFAOYSA-N
disilver(1+) sulfanediide
[S--].[Ag+].[Ag+]
This item is not returnable. View return policy
21548-73-2
Ag2S
247.80
MFCD00003406
XUARKZBEFFVFRG-UHFFFAOYSA-N
disilver(1+) sulfanediide
[S--].[Ag+].[Ag+]

Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 825°C
Color Yellow
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Solubility Information Soluble in HNO3,alkali cyanide solutions. Insoluble in water
Formula Weight 247.8
Odor Odorless
Sensitivity Light sensitive
Appearance Dark gray to black powder/flake/crystalline/beads
Density 7.317 g/cm3 at 20°C
Chemical Name or Material Silver sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'