missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059843
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
21548-73-2
Ag2S
247.80
MFCD00003406
XUARKZBEFFVFRG-UHFFFAOYSA-N
disilver(1+) sulfanediide
[S--].[Ag+].[Ag+]
This item is not returnable. View return policy
21548-73-2
Ag2S
247.80
MFCD00003406
XUARKZBEFFVFRG-UHFFFAOYSA-N
disilver(1+) sulfanediide
[S--].[Ag+].[Ag+]

Silver sulfide is used as photosensitizer in photography, anti-bacterial and anti-microbial agents. It is also useful as qualitative inorganic analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 825°C
Color Gray to Black
Physical Form Pieces
Quantity 5 g
UN Number UN3077
Merck Index 14,8530
Solubility Information Insoluble in water.
Formula Weight 247.8
Percent Purity 99.999%
Odor Odorless
Sensitivity Light sensitive
Density 7.317 g/mL
Assay (metals basis)
Chemical Name or Material Silver sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'