missing translation for 'onlineSavingsMsg'
Learn More

Silver Sulfate Solution 1% (w/v) (d=1.84) In Sulfuric Acid, for COD, Fisher Chemical™

Brand:  Fisher Chemical J/7400/PC15

19.86 GBP valid until 2021-03-31
Use promo code "15537" to get your promotional price.



Code : 23

Additional Details : CAS Number : 10294-26-5 Weight : 1.00000kg Transport : UN number : 1830 Chem class : 8 Pack group : II


Quantity 1L

 View more versions of this product

Product Code. 12626727

  • £123.00 / 1 L

In stock, ready to ship: 24

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

10294-26-5
311.792
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
159865
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
Ag2O4S
3407
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
disilver;sulfate

Specifications

10294-26-5
3407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
disilver;sulfate
159865
Liquid
Colorless
1L
Ag2O4S
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
311.792
311.79g/mol
Plastic Coated Glass bottle
1
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silver sulfate, 10 g/l solution in sulfuric acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents