missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver sulfate, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Silver(I) sulfate is employed in silver plating, and in electrolysis. It is a useful non-staining substitute to silver nitrate. Silver sulfate/iodine is an excellent iodination reagent for alkyl, alkoxybenzenes, aromatic amines and uridines. It is used in batteries; its pharmaceutical composition is used as an antimicrobial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1085°C (decomposition) |
Melting Point | 657°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.999% (metals basis) |
Solubility Information | Partially soluble in water. Water solubility increases with temperature. Soluble in nitric acid,ammonia and conc. sulfuric acid. |
Formula Weight | 311.8 |
Odor | Odorless |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'