missing translation for 'onlineSavingsMsg'
Learn More

Silver Powder, crystalline, -60+80 mesh, Premion™, 99.99% (Metals basis), Thermo Scientific™

Catalog Number p-4824936
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Silver
Appearance Silver-colored powder
Assay Percent Range 99.99% (Metals basis)
Quantity 10 g
Mesh Size -60+80 Mesh
Odor Characteristic
Form Crystalline Powder
Chemical Name or Material Silver
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'