missing translation for 'onlineSavingsMsg'
Learn More

Silver powder, APS 1-3 micron, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048752
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This item is not returnable. View return policy
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3077
Quantity 10 g
Percent Purity 99.9%
Odor Characteristic
Particle Size APS 1 to 3 Micron
Form Powder
Assay (metals basis)
Chemical Name or Material Silver powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'