missing translation for 'onlineSavingsMsg'
Learn More

Silver plate, 3.175mm (0.125in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11306758
Click to view available options
Quantity:
5 x 5 cm
10 x 10 cm
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 x 5 cm
Percent Purity 99.95%
Odor Characteristic
Weight ≈83.3g/5x5cm
Form Plate
Assay (metals basis)
Chemical Name or Material Silver plate, 3.175mm (0.125 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'