missing translation for 'onlineSavingsMsg'
Learn More

Silver plasma standard solution, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 013849.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11311419

  • £55.70 / 100mL


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Silver
Liquid
MFCD00003397
plasma standard solution
Ag in 5% HNO3

Specifications

Silver
Colourless liquid
Ag in 5% HNO3
Nitric acid: 5%; Silver: 0.1%; Water: 94.9%
Colorless
plasma standard solution
Liquid
MFCD00003397
23 hPa (17mm Hg) at 20°C
100mL
Videos
SDS