missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver phosphate, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Silver phosphate finds applications in photography, and in silver staining of biological materials. It is used as a photocatalyst for the visible light photochemical splitting of water, and for production of activated oxygen by the same method. It is employed in Crystal growth for scintillation, optics and dosimetry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 849°C |
Color | Gray to Black |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99% |
Solubility Information | Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3,ammonia,ammonium carbonate,alkali cyanides and thiosulfates. |
Formula Weight | 418.58 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | Yellow powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'