missing translation for 'onlineSavingsMsg'
Learn More

Silver perrhenate, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7059370
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
20654-56-2
AgO4Re
358.071
MFCD00014144
JRISYOCHUNKJAV-UHFFFAOYSA-N
silver i perrhenate, silver perrhenate, rhenium silver tetraoxide, acmc-20ajpz, silver i perrhenate, for synthesis, silver 1+ ion tetraoxorhenate 1-, silver i perrhenate trace metals basis
11089628
silver;oxido(trioxo)rhenium
[O-][Re](=O)(=O)=O.[Ag+]
This item is not returnable. View return policy
20654-56-2
AgO4Re
358.071
MFCD00014144
JRISYOCHUNKJAV-UHFFFAOYSA-N
silver i perrhenate, silver perrhenate, rhenium silver tetraoxide, acmc-20ajpz, silver i perrhenate, for synthesis, silver 1+ ion tetraoxorhenate 1-, silver i perrhenate trace metals basis
11089628
silver;oxido(trioxo)rhenium
[O-][Re](=O)(=O)=O.[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 430°C
Color White
Physical Form Powder
Quantity 1 g
Linear Formula AgReO4
UN Number UN1479
Formula Weight 358.1
Percent Purity 99.99%
Sensitivity Light sensitive
Density 7.05 g/mL
Assay (metals basis)
Chemical Name or Material Silver perrhenate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'